Orders LinkContactUsLinkComponent
Objectives for REBEL Microscopes, ECHO
Objectives for REBEL Microscopes, ECHO
10× Plan Achromat, CSC 0.17, 12 mm, N.A. 0.25
Catalog # 76307-554
Supplier:   DISCOVER ECHO INC.
Supplier #:  RMAG0102
Objectives for REBEL Microscopes, ECHO
Catalog # 76307-554
Supplier:  DISCOVER ECHO INC.
Objectives for REBEL Microscopes, ECHO
10× Plan Achromat, CSC 0.17, 12 mm, N.A. 0.25
Price
Inquire For Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Specifications
  • Description:
    10× Plan Achromat, CSC 0.17
  • Aperture:
    N.A. 0.25
  • Working distance:
    12 mm