Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

RoboSep™ Buffer

Buffer containing FBS, 250 ml
Catalog # MSPP-20104
Marketsource
 :  20104
RoboSep™ Buffer
Catalog # MSPP-20104
Marketsource
RoboSep™ Buffer
RoboSep™ Buffer

Buffer containing FBS, 250 ml

Price
$51.15
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

RoboSep™ buffer is recommended for EasySep™ cell separation protocols performed by RoboSep™.

Please note that one or two bottles of buffer are included with every purchase of a RoboSep™ reagent kit.

Species: Human, mouse, non human primate, rat and others.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications
  • Capacity:
    250 ml
  • Description:
    Buffer containing FBS
  • Application:
    Immunology