Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ASSAY KIT RHOA G-LISA ACTIVATION 24ASSAY
Catalog # MSPP-BK124-S
Supplier:
CYTOSKELETON MS
Expand image
ASSAY KIT RHOA G-LISA ACTIVATION 24ASSAY
Catalog # MSPP-BK124-S
Supplier:
CYTOSKELETON MS
Supplier Number:
BK124-S