Orders LinkContactUsLinkComponent
Polypropylene Stoppers
Polypropylene Stoppers
24/29
Catalog # 470310-648
Supplier:   Wards
Supplier #:  CH0873E
Polypropylene Stoppers
Catalog # 470310-648
Supplier:  Wards
Polypropylene Stoppers
Polypropylene Stoppers
24/29
Price
$5.25
Log in for availability
Each Pack of 10
Product Details & Documents
These lightweight and durable polypropylene stoppers are acid proof and autoclavable.
Specifications
  • Stopper no.:
    24/29