Polypropylene Stoppers
19/26
Catalog # 470310-646
Supplier: Wards
Supplier #: CH0873D
Jump to:
Product Details & Documents
These lightweight and durable polypropylene stoppers are acid proof and autoclavable.
Specifications
- Stopper no.:19/26