Orders LinkContactUsLinkComponent
Polypropylene Stoppers
Polypropylene Stoppers
12/21
Catalog # 470310-642
Supplier:   Wards
Supplier #:  CH0873B
Polypropylene Stoppers
Catalog # 470310-642
Supplier:  Wards
Polypropylene Stoppers
Polypropylene Stoppers
12/21
Price
$4.00
Log in for availability
Each Pack of 10
Product Details & Documents
These lightweight and durable polypropylene stoppers are acid proof and autoclavable.
Specifications
  • Stopper no.:
    12/21