Storage Bins
Stacking bins, Red, PP, Case of 45, 7.56×5.87×4.13"
10117-829
: Treston
: 1520-5
These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.
- Straight sides ensure an efficient use of space
- Multiple color options make finding the right item easy
- Open front provides easy access
- Corrugated base helps retrieve contents, especially smaller sized and flat objects
: Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.
- Color:Red
- Description:Stacking bins
- Dimensions:7.56×5.87×4.13"
- Material:PP
- Size:Case of 45



