Storage Bins
Shelf bins, Red, PP, Case of 20, 19.70×5.2×3.94"
Catalog # 10117-777
Supplier: Treston
: 5015-5
Jump to:
Product Details & Documents
These high quality, durable Treston bins have been designed for daily use in stores, production facilities and warehouses.
- Straight sides ensure an efficient use of space
- Multiple color options make finding the right item easy
- Open front provides easy access
- Corrugated base helps retrieve contents, especially smaller sized and flat objects
: Labels and dividers sold separately, (translucent plastic), labels and blanking piece FP-6M to produce flat surface for adhesive barcode labels on inside rear wall of bins. Contact Avantor representative for more information. Installation is required. Assembly videos available.
Specifications
- Color:Red
- Description:Shelf bins
- Dimensions:19.70×5.2×3.94"
- Material:PP
- Size:Case of 20



