RECOM PD-L1/B7-H1 >95% CARRIER-FR 100 UG
Catalog # 103704-914
Supplier: Novus Biologicals
RECOM PD-L1/B7-H1 >95% CARRIER-FR 100 UG
Catalog # 103704-914
Supplier: Novus Biologicals
Supplier Number: 9326-B7-100
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
Falcon® Cell Strainers, Sterile
Supplier: Corning
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International