Objectives for REVOLVE Microscopes, Echo
60× U-Plan fluorite objective, 0.2 mm, 0.9 NA
Catalog # 76299-624
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0609
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:60×
- Description:60× U-Plan fluorite objective
- Aperture:0.9 NA
- Working distance:0.2 mm