Objectives for REVOLVE Microscopes, Echo
40× U-Plan apochromat oil 340 nm objective, 0.01 mm, 1.35 NA
76299-620
: JMAG0411
- Magnification:40×
- Description:40× U-Plan apochromat oil 340 nm objective
- Aperture:1.35 NA
- Working distance:0.01 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



