Objectives for REVOLVE Microscopes, Echo
10× C plan fluorite phase objective, 9.5 mm, 0.3 NA
Catalog # 76299-618
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0109
Jump to:
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:10×
- Description:10× C plan fluorite phase objective
- Aperture:0.3 NA
- Working distance:9.5 mm