REAGENT COLLAGENASE TYPE IV 1MG/ML 100ML
Catalog # MSPP-07909
Supplier: Stemcell Technologies
REAGENT COLLAGENASE TYPE IV 1MG/ML 100ML
Catalog # MSPP-07909
Supplier: Stemcell Technologies
Supplier Number: 07909
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Premium Aluminum Foil
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International