Anti-CDCP1 Rabbit Polyclonal Antibody
50 µl
Catalog # 103596-148
Jump to:
Discontinued
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Produced in rabbits immunized with purified, recombinant Mouse CDCP1 (rM CDCP1; Catalog#51110-M08H; Q5U462; Met1-Leu666). CDCP1 specific IgG was purified by Mouse CDCP1 affinity chromatography.
Type: Primary
Antigen: CDCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Documents
Specifications
- Size:50 µl
- Clonality:Polyclonal
- Form:Liquid
- Storage Buffer:0.2 μm Filtered solution in PBS
- Isotype:IgG
- Host:Rabbit
- Cat. No.:103596-148
- Reactivity:Mouse
- Shipping Temperature:Ambient
- Antibody Type:Primary
- Antigen Symbol:CDCP1
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:TRASK|SIMA135|CD318
- Antigen Name:CUB domain containing protein 1
- ELISA:Yes



