Orders LinkContactUsLinkComponent
GBS Detect/Strep Reagents
GBS Detect/Strep Reagents
GBS Detect™, for Non-Hemolytic Group B Strep, Reduced Stacking Ring for Automated Streaking Devices, 15×100 mm plate, —
  76288-444
Supplier #:  GA300
GBS Detect/Strep Reagents
  76288-444
 :  
GBS Detect/Strep Reagents
GBS Detect/Strep Reagents
GBS Detect™, for Non-Hemolytic Group B Strep, Reduced Stacking Ring for Automated Streaking Devices, 15×100 mm plate, —
 :  
$113.00
Pack of 10
GBS Detect is used for the isolation and differentiation by enhanced hemolysis of gamma hemolytic Group B Streptococcus.
  • Grouping reagents for the rapid latex agglutination test
  • Lancefield grouping includes A,B,C,D,F,G latex, controls, extraction reagents, cards, and mixing sticks
  • StrepQuick™ to aid in the identification of gram positive cocci
GBS Detect™, for non-hemolytic Group B Strep. Granada Medium for hemolytic Group B Strep. Grouping Reagents A, B, C, and G, for the rapid latex agglutination test. PYR Test Kit with chromogenic developer on paper disks. Selective Beta Strep Agar, for Group A Strep only (Streptococcus pyogenes), especially for throat cultures. Selective Strep Agar (COBA Medium), for the selective isolation of all species of Strep. ci. StrepQuick™ performs a PYR, LAP, and Esculin test to aid in the identification of gram positive coc.
  • Description:
    GBS Detect™, for Non-Hemolytic Group B Strep, Reduced Stacking Ring for Automated Streaking Devices
  • Size:
  • Format:
    15×100 mm plate