Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Laboratory Warning Labels

Warning labels - RADIOACTIVE, Red on yellow, 3", 69×25.4 mm (2.72×1"), 1000 labels/roll (1 label across)
  76276-146
 :  807656026701
Laboratory Warning Labels
  76276-146
Laboratory Warning Labels
Laboratory Warning Labels

Warning labels - RADIOACTIVE, Red on yellow, 3", 69×25.4 mm (2.72×1"), 1000 labels/roll (1 label across)

 
$54.91
   
Roll of 1
 

Various warning and safety labels for use throughout the lab. Designed to inform personnel of the multiple hazards present in the lab, including the presence of radioactive material, cytotoxic compounds, and biological waste.

  • Temperature range: ambient temperature
  • Removable or permanent adhesive
  • All labels are preprinted

Multi-purpose warning labels, ideal for offices, clinics, and laboratories. Paper warning labels feature a permanent adhesive to ensure they won't wrinkle or peel, and are designed for use at ambient temperatures. These general-use safety labels are fully customizable with logos, images, and alphanumeric text, in order to display appropriate warning information.

 
  • Color:
    Red on yellow
  • Description:
    Warning labels - RADIOACTIVE
  • Dimensions:
    69×25.4 mm (2.72×1")
  • Packaging:
    1000 labels/roll (1 label across)
  • Core size:
    3"