Stackable Metal Slide Cabinets, Avantik
6-Drawer Stackable Metal Slide Cabinet, White
Catalog # 76270-350
Supplier: Avantik
Supplier #: ST7076
Jump to:
Product Details & Documents
Holds up to 5000 standard 3×1" microscope slides.
- Interlocking, stackable to 10 cabinets high
- Cabinet and base are stackable with the Boekel Cabinet
- Cabinet base features rubber feet to prevent scratching and slipping
Cabinet is supplied with a two-year warranty.
: Includes plastic drawer dividers, sponge blocks spaces and identification labels.
Specifications
- Color:White
- Description:6-Drawer Stackable Metal Slide Cabinet