Synthware Round-Bottom Flasks with Three Necks, Angled
3000 ml, 45/50, 24/40
Catalog # 76264-090
Supplier: Kemtech America
Supplier #: F43543L
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 76264-090
Supplier: Kemtech America
Jump to:
Product Details & Documents
Round bottom with [ST] joints.
- 3 Angled necks
- Ground glass joints
- Heavy wall Borosilicate construction
- Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Specifications
- Capacity:3000 ml
- Center neck [ST] joint:45/50
- Side neck ST joints:24/40