Orders LinkContactUsLinkComponent
dCTP CF® Dye Conjugates, Biotium
dCTP CF® Dye Conjugates, Biotium
CF®640R dCTP, lyophilized, 642/662 nm, 5 nmol
Catalog # 76221-650
Supplier:   Biotium
Supplier #:  40066-T
dCTP CF® Dye Conjugates, Biotium
Catalog # 76221-650
Supplier:  Biotium
CAS Number:  
dCTP CF® Dye Conjugates, Biotium
CF®640R dCTP, lyophilized, 642/662 nm, 5 nmol
CAS Number:  
Price
$150.10
Log in for availability
Each (5Nmo)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
CF® dye-dCTP can be used to synthesize probes for in situ hybridization, microarray, or to synthesize labeled DNA probes for in situ hybridization and nucleic acid blotting applications.
  • Synthesize exceptionally bright and photostable fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of nine CF® dye colors from green to far-red
  • Biotiums CF® dyes have superior brightness, photostability, and are water-soluble
Caution: NOTE: CF™594 dCTP is not compatible with PCR labeling of DNA probes.
Specifications
Specifications
  • Description:
    CF®640R dCTP, lyophilized
  • Size:
    5 nmol
  • Ex/em:
    642/662 nm