Orders LinkContactUsLinkComponent
dCTP CF® Dye Conjugates, Biotium
CF®532-dCTP, lyophilized, 527/558 nm, 5 nmol
  76221-646
 :   Biotium
 :  40057-T
dCTP CF® Dye Conjugates, Biotium
  76221-646
 :  Biotium
 :  
dCTP CF® Dye Conjugates, Biotium
CF®532-dCTP, lyophilized, 527/558 nm, 5 nmol
 :  
$121.23
Each (5Nmo)
CF® dye-dCTP can be used to synthesize probes for in situ hybridization, microarray, or to synthesize labeled DNA probes for in situ hybridization and nucleic acid blotting applications.
  • Synthesize exceptionally bright and photostable fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of nine CF® dye colors from green to far-red
  • Biotiums CF® dyes have superior brightness, photostability, and are water-soluble
Caution: NOTE: CF™594 dCTP is not compatible with PCR labeling of DNA probes.
  • Description:
    CF®532-dCTP, lyophilized
  • Size:
    5 nmol
  • Ex/em:
    527/558 nm