Mouse IgG Isotype Control
1 mg
Catalog # 103572-504
Marketsource
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
MAB IGG POLYCLONAL INVIVOMAB MS IGG 1MG Catalog #76555-688
Product 76555-688 is similar to 103572-504 but manufacturer and specifications may vary.
MS PAB TO MOUSE IGG Catalog #89352-766
Product 89352-766 is similar to 103572-504 but manufacturer and specifications may vary.
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
InVivoMab polyclonal mouse IgG, Clone: Polyclonal, Isotype: mouse IgG, Applications: in vivo administration
- Recommended Dilution Buffer: IP0070
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Specifications
- Size:1 mg
- Cat. No.:103572-504
- Antigen Symbol:IgG
- Host:Mouse



