VWR® Basic Industrial Lab Coats
Product Details & Documents
Disposable lab coat made from 20 gram spunbond polypropylene (SPP) fabric. Provides lightweight, breathable protection for non-hazardous environments. Ideal for general industrial use, food processing, and clean areas requiring basic contamination control.
Lightweight and breathable fabric reduces heat buildup during extended use
Snap front closure for easy donning and removal
- Elastic wrists and ankles for better coverage
- Single-use, disposable design for contamination control
- Economical solution for basic protection needs
- Non-linting material helps maintain clean environments
- Suitable for general industrial, food processing, and non-hazardous environments
The VWR® basic protection lab coat is designed to provide reliable, economical protection in environments where contamination control is essential but exposure risks are minimal. Constructed from 20 gram spunbond polypropylene (SPP), this garment offers a lightweight and breathable solution that ensures user comfort during extended wear. The non-woven SPP fabric is non-linting, reducing the risk of particle contamination, and is suitable for clean areas, food processing, and general industrial applications.
Key design features include a snap front closure for easy donning and removal, and an elastic cuff option is also available. The disposable nature of the garment supports hygiene compliance.
: Lab coats are packaged 30 per case.
Specifications
- Clothing Size:3XL
- Color:Blue
- Length:45½"
- Chest size:28½"
- Sleeve:23"
- Cat. No.:76218-718
- Sleeve or leg length:Long
- Cleanroom Class:10000/ISO 7
- Garment application:Cleanroom|Industrial|Laboratory
- Gender:Unisex
- Pockets:With Pockets
- Closure type:With Press-Stud Fastening
- Fabric:Spunbound polypropylene
- Particle shedding (Helmke Drum):Level II
- Weight:20 g/m²
- H₂O Resistance:First line barrier
Complementary Products
Product Information
- Catalog #DescriptionSizeAvailabilityPrice



