Various Replacement Parts, Sciencix
Tubing D-L Assembly, LC-30AD, SHIM, 228-53300-41
Catalog # 103572-346
Marketsource
Supplier: SCIENCIX
Supplier #: 11-4152
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Tubing D-L Assembly
- OEM manufacturer:SHIM
- OEM equivalent:228-53300-41
- Model:LC-30AD