Various Replacement Parts, Sciencix
Compression Screws and Ferrules, 1/16", SS, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, Alliance, WATE, WAT025604
103572-340
Marketsource
: SCIENCIX
Supplier #: CTS-21012
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
- Description:Compression Screws and Ferrules, 1/16", SS
- OEM manufacturer:WATE
- OEM equivalent:WAT025604
- Model:717, 2690, 2690D, 2695, 2695D, 2790, 2795, Alliance