Orders LinkContactUsLinkComponent
VWR® Microfiber String Mop
VWR® Microfiber String Mop
Microfiber string mop, irradiated, 16 oz.
Catalog # 76214-292
Supplier:   VWR International
VWR® Microfiber String Mop
Catalog # 76214-292
Supplier:  VWR International
VWR® Microfiber String Mop
VWR® Microfiber String Mop
Microfiber string mop, irradiated, 16 oz.
Price
$1,048.93
Log in for availability
Case of 24
Product Details & Documents
This Microfiber String Mop is an absorbent and hard wearing mop for cleaning and liquid pick-up in controlled and critical environments.
  • Highly absorbent
  • Durable
The unique ribbing of the microfiber strands produces a mop that is both low linting and absorbent. The strands are comprised of split fiber polyester filaments, 30 times smaller than cotton fibers. These microfibers work to pick up tiny particles where cotton/rayon blends and polyesters may often roll over the dirt. The solid constructions of the mop, combined with the durable microfiber, allow the mop to be used with strong disinfectants, laundered and re-used.
Specifications
  • Description:
    Microfiber string mop, irradiated
  • Weight:
    16 oz.