Orders LinkContactUsLinkComponent
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Green
Catalog # 76214-234
Supplier:   Simport Scientific
Supplier #:  M509-4T
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Catalog # 76214-234
Supplier:  Simport Scientific
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Slimsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Green
Price
$1,441.23
Log in for availability
Case of 2,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Slimsette™ Tissue Processing/Embedding Cassette, in Quickload™ stacks.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Certifications: CE-IVD.
Packaging: Each case contains 50 Quickload™ stacks of 40 cassettes.
Specifications
  • Color:
    Green