Orders LinkContactUsLinkComponent
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Green
Catalog # 76214-196
Supplier:   Simport Scientific
Supplier #:  M505-4T
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Catalog # 76214-196
Supplier:  Simport Scientific
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Unisette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Green
Price
$421.02
Log in for availability
Case of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Unisette™ Tissue Processing/Embedding Cassette.
  • In Quickload™ stacks
  • 35° Anterior writing area
  • One-piece construction with hinge-lock design prevents early separation of base and lid
  • Snap latch and hinge-lock design; cassettes will always relock securely
  • Allows for one-hand operation
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
The snap latch and hinge-lock design allows for one-hand operation and always relocks securely without danger of specimen loss.
Certifications: CE-IVD.
Packaging: Each case contains 25 Quickload™ stacks of 40 cassettes.
Specifications
  • Color:
    Green