Orders LinkContactUsLinkComponent
Swingsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
White
Catalog # 76214-150
Supplier:   Simport Scientific
 :  M517-2T
Swingsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Catalog # 76214-150
Supplier:  Simport Scientific
Swingsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
Swingsette™ Tissue Processing/Embedding Cassette in Quickload™ Stacks, Simport
White
Price
$791.60
Log in for availability
Case of 2,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Secure specimens during processing with the Swingsette™ Tissue Processing/Embedding Cassette, in Quickload™ stacks.
  • In Quickload™ stacks
  • 45° anterior writing area
  • Easy to assemble hinged lids
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
 : CE-IVD.
 : Each case contains 50 Quickload™ stacks of 40 cassettes, lids packaged separately.
Specifications
  • Color:
    White