ESD SMD Boxes, Transforming Technologies
Medium, 37×12×15 mm (int)
Catalog # 76209-948
Supplier: Transforming Technologies
: SM0874
Jump to:
Product Details & Documents
Static-safe SMD boxes with spring-loaded hinges are designed to interlock for easy organization and storage.
- Ideal for engineering labs
- Available with conductive black or clear metalized lids
SMD plastic boxes have a unique interlocking modular design. Spring-loaded lids have a stay-open feature for convenient loading and removal of parts. Surface resistivity of 10² to 10⁵ ohms/cm². They are ideal for engineering labs, kitting, in-process handling, and safe transport of static-sensitive components.
: Meets or exceeds requirements of ANSI ESD-S20.20
Specifications
- Description:Medium
- Ext. dimensions:37×12×15 mm (int)