VWR® Covers
VWR® cover for media bottles and flasks, Purple, 7" bulk packaging
Catalog # 76205-184
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Covers provide a convenient means to cover and protect bowls, flanges, flasks, beakers, containers, media bottles, and hoses
- Breathable
- High-quality construction
- Form fitting
- Variety of sizes
- Made in the USA
Covers utilize top materials such SMS, Alliance®, and Tyvek®, which offer a variety of beneficial characteristics.
Ordering information: If the size and material combination required is not listed, simply contact your Avantor Controlled Environment Specialist to assist.
Documents
Certificates
Search by Lot NumberCoQ VWR® Covers for media bottles and flasks.pdf
Conformity declaration
Specifications
- Color:Purple
- Description:VWR® cover for media bottles and flasks
- Fits:7" bulk packaging