Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Multiple Affinity Removal Column - Ms-3 (Mouse), 4.60 mm, 50 mm
Catalog # 76204-858
Supplier: Agilent Technologies
Supplier #: 5188-5217
Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Catalog # 76204-858
Supplier: Agilent Technologies
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Multiple Affinity Removal LC Column – Mouse 3 is designed as a simple way to remove THREE interfering high-abundant proteins from mouse plasma samples.
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
Caution: For research use only.
Documents
Specifications
- Ø int.:4.60 mm
- Description:Multiple Affinity Removal Column - Ms-3 (Mouse)
- Length:50 mm



