ANTIBODY NEUN FOX 3/RBFOX 3 100UL
Catalog # MSPP-266011SY
Supplier: Cedar Lane
ANTIBODY NEUN FOX 3/RBFOX 3 100UL
Catalog # MSPP-266011SY
Supplier: Cedar Lane
Supplier Number: 266011(SY)
Frequently Bought Together
Static Mouse Cage, 1284L
Supplier: Tecniplast Usa
Probe Mall
Supplier: DR INSTRUMENTS INC. SE
PVC Film Wrap, T FRANK MCCALLS INC
Supplier: TFrank McCalls
Reusable Plastic Animal Cages, Tecniplast®
Supplier: Tecniplast Usa
VWR® Shelf Liners
Supplier: VWR International
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Supplier: Thermo Fisher Scientific
Biodyne™ B Nylon Membrane, 0.45 µm, Thermo Scientific
Supplier: Invitrogen
PADS LIGHT DUTY CLEANING SYSTEM 10IN
Supplier: ULINE MS
UNDERPAD HIGH-ABSORBANCE 23X36IN CS72
Supplier: COVETRUS NORTH AMERICA MS
Swab, Foam Tip, Sterile
Supplier: Hardy Diagnostics
neodisher® Detergents, Miele
Supplier: Miele
COVER SHOE STANDARD SZ 12-15 CS150
Supplier: ULINE MS
SUTURE V20 2-0 15CM CS12
Supplier: CIA MEDICAL MS
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Human Recombinant Serpin A3/alpha 1-Antichymotrypsin (from NS0 Cells)
Supplier: R&D Systems
SUTURE ABSORBABLE 3-0 / C-14 / 30IN
Supplier: COVETRUS NORTH AMERICA MS