Demonstration Pulleys
Demonstration Pulleys, Double
Catalog # 470235-698
Supplier: GSC International
Supplier #: 4218-00-10
Jump to:
Product Details & Documents
Jumbo-Sized Pulleys are Designed for Classroom Demonstrations
These pulleys are open to make it easier for users to run pulley cords through them. The single, double, and triple pulleys are all 2 3/4" in diameter. Light-weight, low-friction, and deformation-resistant plastic ensures accurate results.
Specifications
- Description:Demonstration Pulleys, Double