Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ASSAY KIT RAS G-LISA ACTIVATION 96 TESTS
Catalog # MSPP-BK131
Supplier:
CYTOSKELETON MS
Expand image
ASSAY KIT RAS G-LISA ACTIVATION 96 TESTS
Catalog # MSPP-BK131
Supplier:
CYTOSKELETON MS
Supplier Number:
BK131