Wringers, Contec®
Flat Mop Wringer, Stainless steel, VertiKlean® MAX™ and QuickTask™ Mops (15 l (3.9 gal.) and 25 l (6.5 gal.) Buckets)
Supplier: CONTEC, INC. MX
: 2689
Jump to:
Product Details & Documents
A variety of wringers designed for use with curved and flat mops.
- Stainless steel and polypropylene wringers for a variety of Contec® bucket systems
- Can be used with curved, flat, and string mops
- Fully autoclavable
These wringers fit on Contec® 3.9 gallon (15 l), 5 gallon (20 l) , and 6.5 gallon (25 l) bucket systems. All wringers are compatible with a wide variety of disinfectants.
Specifications
- For Use With:VertiKlean® MAX™ and QuickTask™ Mops (15 l (3.9 gal.) and 25 l (6.5 gal.) Buckets)
- Description:Flat Mop Wringer
- Material:Stainless steel
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement spring, electropolished stainless steel, for Downpress Flat Mop Wringer 2756Replacement spring, electropolished stainless steel, for Downpress Flat Mop Wringer 275676519-522Log in for availabilityEach - $20.10