CELLS MACROPHAGE BONE MARROW LADMAC
Catalog # MSPP-CRL-2420
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
CELLS MACROPHAGE BONE MARROW LADMAC
Catalog # MSPP-CRL-2420
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: CRL-2420
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Corning® Trypsin
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
Parafilm® M Sealing Film
Supplier: Heathrow Scientific
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
ATCC® RPMI-1640 Medium, American Type Culture Collection
Supplier: AMERICAN TYPE CULTURE COLLECTION MS