Reagent Troughs
Disposable Trough Liner, PP, 100 ml
Catalog # 76197-168
Supplier: TWD Scientific
Supplier #: CT-L-100
Jump to:
Product Details & Documents
Reagent Troughs are made of polypropylene. Troughs support a disposable polypropylene trough liner. Liners are made of thin, disposable, lightweight polypropylene.
- V-shaped bottom and angled sides for maximum reagent recovery
- Usable with up to 12-channel pipettors
- Designed for SBS format applications and compatible with robotic systems
- Reusable
Perfect for 12 or 8-channel pipettes and compatible eith robotic systems.
Specifications
- Capacity:100 ml
- Description:Disposable Trough Liner
- Material:PP