Lab Scoops
Lab Scoop, Stainless steel, 6 - 10 mg
Catalog # 76197-158
Supplier: TWD Scientific
Supplier #: GSS-03-25
Jump to:
Product Details & Documents
Disposable Lab Scoops allow for easy and rapid transfer of milligram quantities. Scoops are available in either anti-static polypropylene or stainless steel construction.
- Cost effective
- Reduces time and labor of repetitive weighing
- Easy dispense packages
These cost-effective scoops are great for rapid transfer of milligram quanities.
Specifications
- Capacity:6 - 10 mg
- Description:Lab Scoop
- Material:Stainless steel