Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
CRITERION™ Phosphate Buffer, pH 7.2

CRITERION™ Phosphate Buffer, pH 7.2

10 kg, Bucket
Catalog # 76196-446
Product available on GSA Advantage®
Supplier:   Hardy Diagnostics
Supplier #:  C8333
CRITERION™ Phosphate Buffer, pH 7.2
Catalog # 76196-446
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
CRITERION™ Phosphate Buffer, pH 7.2
CRITERION™ Phosphate Buffer, pH 7.2

10 kg, Bucket

CAS Number:  
Price
$1,490.00
Log in for availability
Each (10kg)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CRITERION™ Agar, bacteriological grade is used as a solidifying agent in the preparation of solid and semi-solid culture media.

  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • Increased solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • BSE-free
Specifications

Specifications

  • Pack type:
    Bucket
  • Size:
    10 kg
  • Application:
    Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
  • Cat. No.:
    76196-446
  • Storage Temperature:
    2...30 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.