Multiple Affinity Removal Column Human 14, Agilent Technologies
Multiple Affinity Removal Column, Human-14, 4.60 mm, 50 mm
Catalog # 76193-736
Supplier: Agilent Technologies
Supplier #: 5188-6557
Multiple Affinity Removal Column Human 14, Agilent Technologies
Catalog # 76193-736
Supplier: Agilent Technologies
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The Human 14 Multiple Affinity Removal Column is designed to chromatographically remove fourteen interfering high-abundant proteins from human plasma samples.
- Made of PEEK (polyetheretherketone)
- High-capacity serum load
- Affinity resin packed
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
Documents
Specifications
- Ø int.:4.60 mm
- Description:Multiple Affinity Removal Column, Human-14
- Length:50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



