Liquid Antiserums
Anti-Rho (D) 5 mL bottle, 50
Catalog # 470233-570
Supplier: Ward's Science
Supplier #: IS3152
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Liquid antiserums are pooled and ready to use.
- Anti-A and B Serums Identify ABO Blood Group
- Anti-Rho (D) Identifies + or - Rh Factor
- No Refrigeration Necessary
- Pooled
- Ready-To-Use
Anti-A and B serums identify ABO blood groups (A, B, AB, or O), and anti-AB provides a check on type O determinations. Anti-Rho (D) identifies + or - Rh factor.
Refrigeration is not necessary. The antiserums are contained in dropper bottles. They are for educational purposes only.
Specifications
- Description:Anti-Rho (D) 5 mL bottle
- No. of students:50