Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dulbecco's Modified Eagle's Medium (DMEM), Quality Biological

Dulbecco's Modified Eagle's Medium (DMEM), Quality Biological

DMEM without Sodium pyruvate and L-Glutamine, Liquid, 12 Months, EA, 500 ml
Catalog # 76176-672
   Sustainable Packaging
Supplier #:  112-034-101
Dulbecco's Modified Eagle's Medium (DMEM), Quality Biological
Catalog # 76176-672
   Sustainable Packaging
CAS Number:  
Dulbecco's Modified Eagle's Medium (DMEM), Quality Biological
Dulbecco's Modified Eagle's Medium (DMEM), Quality Biological

DMEM without Sodium pyruvate and L-Glutamine, Liquid, 12 Months, EA, 500 ml

CAS Number:  
Price
$85.71
Log in for availability
Each (500ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

DMEM (Dulbecco’s Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells. We offer a variety of DMEM modifications for a range of cell culture applications.

  • With: High Glucose
  • Without: HEPES
  • Endotoxin and Mycoplasma tested

DMEM is one of the most widely used modifications of Basal Medium Eagle (BME). DMEM was originally developed as a richer version of BME; with 4 times higher concentrations of amino acids, vitamins and some additional components than BME. DMEM is commonly used for the culture of normal and transformed cells.

Recommendation
To DMEM without L-glutamine, it is recommended to add L-glutamine prior to use at a final concentration of 4.0 mM.

DMEM with high glucose supports increased cell growth. DMEM without phenol red is beneficial to downstream purification processing of expressed proteins (i.e. mAb, VLP) that require the use of various chromatography processes. The addition of sodium bicarbonate creates a high buffer capacity in the medium and keeps the pH-value in the physiological area during cultivation. DMEM: Ham's F-12 without L-glutamine contains no proteins, lipids or growth factors; and uses a sodium bicarbonate system (1.2 g/L), therefore requiring a 5 to 10% CO2 environment to maintain physiological pH. L-glutamine addition, to DMEM without L-glutamine, at the time of use allows for limited degradation over time.

QBI offers a variety of DMEM modifications for a range of cell culture applications.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Caution: Research Use Only

Specifications

Specifications

  • Media Format:
    Liquid
  • Pack type:
    EA
  • Description:
    DMEM without Sodium pyruvate and L-Glutamine
  • Size:
    500 ml
  • Shelf life:
    12 Months
  • Cat. No.:
    76176-672
  • Environmental Certification:
    ECO LOGOS
  • Environmentally Preferable:
    Sustainable Packaging
  • Sterility:
    Sterile filtered
  • Shipping Conditions:
    Ambient temperature

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.