VWR® Covers
VWR® cover for media bottles and flasks, White, 3" bulk packaging
76176-630
VWR® Covers provide a convenient means to cover and protect bowls, flanges, flasks, beakers, containers, media bottles, and hoses
- Breathable
- High-quality construction
- Form fitting
- Variety of sizes
- Made in the USA
Covers utilize top materials such SMS, Alliance®, and Tyvek®, which offer a variety of beneficial characteristics.
: If the size and material combination required is not listed, simply contact your Avantor Controlled Environment Specialist to assist.
- Color:White
- Description:VWR® cover for media bottles and flasks
- Fits:3" bulk packaging