Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STANDARD CANNABICHROMENE 1000 UG/ML 1 ML
Catalog # 76124-770
Supplier:
Restek
SDS
Certificates
Expand image
STANDARD CANNABICHROMENE 1000 UG/ML 1 ML
Catalog # 76124-770
Supplier:
Restek
Supplier Number:
34092
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_34092
Documentation