CELL LINE NCI-H345 [H345]
Catalog # MSPP-HTB180
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
CELL LINE NCI-H345 [H345]
Catalog # MSPP-HTB180
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: HTB-180
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Heathrow Scientific
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek