Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Oxoid CampyGen Compact

CampyGen plastic pouches, 6×13", 20 per box
Catalog # 76076-688
Supplier:   Hardy Diagnostics
 :  AG020C
Oxoid CampyGen Compact
Catalog # 76076-688
Supplier:  Hardy Diagnostics
Oxoid CampyGen Compact
Oxoid CampyGen Compact

CampyGen plastic pouches, 6×13", 20 per box

Price
$121.00
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.

The Oxoid CampyGen and CampyGen Compact are simple-to-use systems for the rapid generation of a microaerophilic environment necessary to cultivate Campylobacter species.

Like the AnaeroGen, the CampyGen also requires no catalyst and no water activation steps. No dangerous gas is produced. The final atmosphere is reduced to 6% oxygen, and supplemented with 14% carbon dioxide which is ideal for growth of Campylobacter and other microaerophilic organisms. Sachet, bags and clips are sold separately.

Ordering information: Pouches and clips sold separately.

Specifications
  • Description:
    CampyGen plastic pouches
  • Size:
    6×13", 20 per box
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.