Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Hyper Microtubes, Watson Bio Lab

Hyper Microtubes, Watson Bio Lab

Microtube with Slight Stiff-Touch Cap, 2.0 ml
Catalog # 75982-891
Supplier:   Watson Bio Lab
Supplier #:  132-620C
Hyper Microtubes, Watson Bio Lab
Catalog # 75982-891
Supplier:  Watson Bio Lab
Hyper Microtubes, Watson Bio Lab
Hyper Microtubes, Watson Bio Lab

Microtube with Slight Stiff-Touch Cap, 2.0 ml

Price
$133.29
Log in for availability
Pack of 1,000
Product Details & Documents

These hyper microtubes feature a writing space, a locking mechanism, and a friendly hinge for fingers.

  • Autoclavable Polypropylene
  • Writing Space
  • Locking Mechanism
  • Hinge for Fingers
  • Scale Graduations

The flat top allows for marking on the microtubes' top, and the surface of the tube itself offers more writing space.

The round-bottom microtubes feature autoclavable polypropylene construction. A scale marks each 0.5 ml on the tubes. The lock part prevents the thumb from touching the inner cap.

 : Pestles, storage racks, and cap locks are available separately.

Specifications
  • Volume:
    2.0 ml
  • Description:
    Microtube with Slight Stiff-Touch Cap