Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Hyper Microtubes, Watson Bio Lab

Hyper Microtubes, Watson Bio Lab

Microtube with Soft-Touch Cap, 1.5 ml
Catalog # 75982-879
Supplier:   Watson Bio Lab
Supplier #:  131-7155C
Hyper Microtubes, Watson Bio Lab
Catalog # 75982-879
Supplier:  Watson Bio Lab
Hyper Microtubes, Watson Bio Lab
Hyper Microtubes, Watson Bio Lab

Microtube with Soft-Touch Cap, 1.5 ml

Price
$92.51
Log in for availability
Pack of 1,000
Product Details & Documents

These hyper microtubes feature a writing space, a locking mechanism, and a friendly hinge for fingers.

  • Autoclavable Polypropylene
  • Writing Space
  • Locking Mechanism
  • Hinge for Fingers
  • Scale Graduations

The flat top allows for marking on the microtubes' top, and the surface of the tube itself offers more writing space.

The round-bottom microtubes feature autoclavable polypropylene construction. A scale marks each 0.5 ml on the tubes. The lock part prevents the thumb from touching the inner cap.

Accessories information: Pestles, storage racks, and cap locks are available separately.

Specifications
  • Volume:
    1.5 ml
  • Description:
    Microtube with Soft-Touch Cap
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.