Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sequencng Binding Buffer, Zymo Research

Sequencng Binding Buffer, Zymo Research

500 mL
Catalog # 77001-312
Supplier:   Zymo Research
Supplier #:  D4050-1-500
Sequencng Binding Buffer, Zymo Research
Catalog # 77001-312
Supplier:  Zymo Research
CAS Number:  
Sequencng Binding Buffer, Zymo Research
Sequencng Binding Buffer, Zymo Research

500 mL

CAS Number:  
Price
$456.39
Log in for availability
Each (500ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Specifications

Specifications

  • Size:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.