Orders LinkContactUsLinkComponent
VWR® Glove Liners
VWR® Glove Liners
Catalog # 76034-602
VWR® Glove Liners
Catalog # 76034-602
Supplier:  
VWR® Glove Liners
VWR® Glove Liners
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
GLOVELINER SK FF NYLN SM CS200 Catalog #14444-316
Product 14444-316 is similar to 76034-602 but manufacturer and specifications may vary.
Product Details & Documents
Ergonomically designed glove liners wick away moisture for comfortable use with disposable gloves.
  • Ambidextrous glove liners
  • Reduces perspiration and wicks away moisture
  • Low-lint, 100% 15 denier nylon composition
  • Sturdy, launderable, and reusable
  • Color-coded cuffs indicate size
  • Pre-laundered; will not shrink
Liners provide protection and comfort when used with latex, nitrile, and PVC gloves. Fully launderable for reusability and economy.
Packaging: Glove Liners are packaged 20 pair per bag, 15 bags per case. They are packed in clean polyethylene bags that are free of silicone, dioctyl phthalate (DOP), and amides.